Learn More
YMC America, Inc ProC18 RS 5um 150x.075mm 1/16
SDP
Catalog No. 5000878012
Encompass
YMC-Pack Pro C18 RS exhibits elevated recognition of stereoselectivity due to high carbon trifunctional bonding Columns maintain a strong resistance to acid and alkali and the separation selectivity for compounds that differ only slightly in hydrophobicity is outstanding In addition extensive endcapping of the stationary phase results in excellent peak shape for basic compounds
Catalog No. 50-008-78012 Supplier YMC America, Inc Supplier No. RS08S0515E8AU
YMC-Pack Pro C18 RS exhibits elevated recognition of stereoselectivity due to high carbon trifunctional bonding Columns maintain a strong resistance to acid and alkali and the separation selectivity for compounds that differ only slightly in hydrophobicity is outstanding In addition extensive endcapping of the stationary phase results in excellent peak shape for basic compounds