Learn More
YMC America, Inc ProC18 RS guard 80A 3um 10x3mm
SDP
Catalog No. 5000879407
Encompass
YMC-Pack Pro C18 RS exhibits elevated recognition of stereoselectivity due to high carbon trifunctional bonding Columns maintain a strong resistance to acid and alkali and the separation selectivity for compounds that differ only slightly in hydrophobicity is outstanding In addition extensive endcapping of the stationary phase results in excellent peak shape for basic compounds
Catalog No. 50-008-79407 Supplier YMC America, Inc Supplier No. RS08S030103GC
YMC-Pack Pro C18 RS exhibits elevated recognition of stereoselectivity due to high carbon trifunctional bonding Columns maintain a strong resistance to acid and alkali and the separation selectivity for compounds that differ only slightly in hydrophobicity is outstanding In addition extensive endcapping of the stationary phase results in excellent peak shape for basic compounds