Learn More
YMC America, Inc ProC4 120A 5um 5x0.5mm 1/16
SDP
Catalog No. 5000877892
Encompass
YMC-Pack C4 is ideal for analyzing mixtures with a wide range of component polarity This is due to the C4 bonded phase exhibiting shorter analysis time while still maintaining high resolution when compared to longer-chain bonded chemistries YMC-Pack C4 (Butyl) columns are available with three pore sizes to accommodate optimum separations of virtually any molecular-weight compound The 120 phase is used extensively with non-polar peptides which are too strongly retained on C8 and C18 The 200 and 300 phase columns effectively resolve many classes of proteins and biopolymers The wide pore butyl chemistry allows minimal distortion of tertiary conformation for large biomolecules and results in samples that are pure concentrated and high in biological activity
Catalog No. 50-008-77892 Supplier YMC America, Inc Supplier No. BS12S05E5J0AU
YMC-Pack C4 is ideal for analyzing mixtures with a wide range of component polarity This is due to the C4 bonded phase exhibiting shorter analysis time while still maintaining high resolution when compared to longer-chain bonded chemistries YMC-Pack C4 (Butyl) columns are available with three pore sizes to accommodate optimum separations of virtually any molecular-weight compound The 120 phase is used extensively with non-polar peptides which are too strongly retained on C8 and C18 The 200 and 300 phase columns effectively resolve many classes of proteins and biopolymers The wide pore butyl chemistry allows minimal distortion of tertiary conformation for large biomolecules and results in samples that are pure concentrated and high in biological activity