Learn More
YMC America, Inc ProC8 guard 120A 5um 20x3mm
SDP
Catalog No. 5000878597
Encompass
YMC-Pack Pro C8 columns are made using advanced endcapping technology to reduce residual silanol activity This results in excellent peak shapes for basic compounds In addition the ultra pure silica and very low residual silanol activity means that these columns frequently do not require the use of ion-pairing reagents or ion-suppression techniques making them ideal for LC/MS applications Surface hydrophobicity is lower than that of ODS making Pro C8 useful for fast analysis of compounds of greatly differing hydrophobic properties Pro C8 can also be useful for separation of hydrophilic and planar compounds in cases where resolution is hard to achieve on ODS materials
Catalog No. 50-008-78597 Supplier YMC America, Inc Supplier No. OS12S050203GC
YMC-Pack Pro C8 columns are made using advanced endcapping technology to reduce residual silanol activity This results in excellent peak shapes for basic compounds In addition the ultra pure silica and very low residual silanol activity means that these columns frequently do not require the use of ion-pairing reagents or ion-suppression techniques making them ideal for LC/MS applications Surface hydrophobicity is lower than that of ODS making Pro C8 useful for fast analysis of compounds of greatly differing hydrophobic properties Pro C8 can also be useful for separation of hydrophilic and planar compounds in cases where resolution is hard to achieve on ODS materials