Learn More
Aviva Systems Biology Corp PROCA1 Antibody - C-terminal region (ARP52989_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PROCA1. It was validated on Western Blot. . Alternate names: MGC39650ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-285-792