Learn More
Aviva Systems Biology Corp PROCA1 Antibody - middle region (ARP52990_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52990_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PROCA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PROCA1. Alternate names: MGC39650ApplicationWB. Species reactivity: Horse, Human, Pig, Rabbit
Catalog No. 50-014-60891