Learn More
TARGETMOL CHEMICALS INC PROCAINAMIDE 50MG
SDP
Catalog No. 502579626
Encompass_Preferred
Also available in 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Procainamide (Novocainamide) is a specific and potent DNA methyltransferase 1 ( DNMT1 ) inhibitor. Procainamide is a Class 1A antiarrhythmic agent. Procainamide has the potential for the research of cancer and arrhythmias [1] [2]. purity: 98%
Catalog No. 50-257-9626 Supplier TARGETMOL CHEMICALS INC Supplier No. T6032250MG
Also available in 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Procainamide (Novocainamide) is a specific and potent DNA methyltransferase 1 ( DNMT1 ) inhibitor. Procainamide is a Class 1A antiarrhythmic agent. Procainamide has the potential for the research of cancer and arrhythmias [1] [2]. purity: 98%