Learn More
QA Bio Inc LudgerClean Procainamide clean-up plate
Catalog No. 502236705
Encompass
Each pack contains one hydrophilic interaction polypropylene 96-well plate for removal of excess procainamide label after procainamide labeling of glycans.
This item is not returnable. View return policy
Catalog No. 50-223-6705 Supplier QA Bio Inc Supplier No. LCPROC96
Each pack contains one hydrophilic interaction polypropylene 96-well plate for removal of excess procainamide label after procainamide labeling of glycans.