Learn More
Apexbio Technology LLC Procainamide HCl 614-39-1 50mg
SDP
Catalog No. 502240627
Encompass_Preferred
Procainamide hydrochloride (CAS 614-39-1) is a small molecule that functions as an inhibitor of DNA methyltransferase 1 (DNMT1) modulating DNA methylation patterns and thereby restoring the expression of tumor suppressor genes This activity has been associated with suppression of cell proliferation and migration as well as the induction of cellular vacuolization Additionally Procainamide hydrochloride has been shown to enhance potassium channel activity contributing to the regulation of smooth muscle relaxation It is commonly employed in biomedical research focused on epigenetic mechanisms in oncology and studies of cardiac arrhythmias
This item is not returnable. View return policy
Catalog No. 50-224-0627 Supplier Apexbio Technology LLC Supplier No. B479850
Procainamide hydrochloride (CAS 614-39-1) is a small molecule that functions as an inhibitor of DNA methyltransferase 1 (DNMT1) modulating DNA methylation patterns and thereby restoring the expression of tumor suppressor genes This activity has been associated with suppression of cell proliferation and migration as well as the induction of cellular vacuolization Additionally Procainamide hydrochloride has been shown to enhance potassium channel activity contributing to the regulation of smooth muscle relaxation It is commonly employed in biomedical research focused on epigenetic mechanisms in oncology and studies of cardiac arrhythmias