Learn More
Apexbio Technology LLC Procainamide HCl 614-39-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000087971
Encompass_Preferred
Procainamide hydrochloride (CAS 614-39-1) is a small molecule that functions as an inhibitor of DNA methyltransferase 1 (DNMT1) modulating DNA methylation patterns and thereby restoring the expression of tumor suppressor genes This activity has been associated with suppression of cell proliferation and migration as well as the induction of cellular vacuolization Additionally Procainamide hydrochloride has been shown to enhance potassium channel activity contributing to the regulation of smooth muscle relaxation It is commonly employed in biomedical research focused on epigenetic mechanisms in oncology and studies of cardiac arrhythmias
Catalog No. 50-000-87971 Supplier Apexbio Technology LLC Supplier No. B47985.1
Procainamide hydrochloride (CAS 614-39-1) is a small molecule that functions as an inhibitor of DNA methyltransferase 1 (DNMT1) modulating DNA methylation patterns and thereby restoring the expression of tumor suppressor genes This activity has been associated with suppression of cell proliferation and migration as well as the induction of cellular vacuolization Additionally Procainamide hydrochloride has been shown to enhance potassium channel activity contributing to the regulation of smooth muscle relaxation It is commonly employed in biomedical research focused on epigenetic mechanisms in oncology and studies of cardiac arrhythmias