Learn More
Medchemexpress LLC Procainamide hydrochloride | 614-39-1 | 99.5% | C13H22ClN3O | 1 mL
SDP
Catalog No. 5000432054
Encompass_Preferred
Procainamide hydrochloride is a specific and potent inhibitor of DNA methyltransferase 1 (DNMT1). It reactivates the expression of tumor suppressor factors by demethylating tumor suppressor genes, and can induce vacuolization in various cell types, reducing cell proliferation and migration. It also relaxes airway smooth muscle by activating potassium channels. This product is suitable for use in cancer and arrhythmia research.
  • Specific and potent inhibitor of DNA methyltransferase 1
  • Reactivates tumor suppressor gene expression
  • Induces cellular vacuolization
  • Reduces cell proliferation and migration
  • Relaxes airway smooth muscle
  • Causes loss of CpG methylation
  • Demethylates ER, p16, and RAR genes
  • Useful in cancer research
  • Useful in arrhythmia research
Catalog No. 50-004-32054 Supplier Medchemexpress LLC Supplier No. HYA008410MM/1ML
Procainamide hydrochloride is a specific and potent inhibitor of DNA methyltransferase 1 (DNMT1). It reactivates the expression of tumor suppressor factors by demethylating tumor suppressor genes, and can induce vacuolization in various cell types, reducing cell proliferation and migration. It also relaxes airway smooth muscle by activating potassium channels. This product is suitable for use in cancer and arrhythmia research.
  • Specific and potent inhibitor of DNA methyltransferase 1
  • Reactivates tumor suppressor gene expression
  • Induces cellular vacuolization
  • Reduces cell proliferation and migration
  • Relaxes airway smooth muscle
  • Causes loss of CpG methylation
  • Demethylates ER, p16, and RAR genes
  • Useful in cancer research
  • Useful in arrhythmia research