Learn More
Abcam Procainamide hydrochloride, Sodium channel blocker, 10G
Catalog No. 502707092
Encompass_Preferred
MW 271.79 g/mol, Purity >99%. Sodium channel blocker. Specific DNA methyltransferase 1 inhibitor (Ki = 7.2 μM). Antiarrhythmic in vivo. Limited ability to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7092 Supplier Abcam Supplier No. AB12095510G
MW 271.79 g/mol, Purity >99%. Sodium channel blocker. Specific DNA methyltransferase 1 inhibitor (Ki = 7.2 μM). Antiarrhythmic in vivo. Limited ability to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement