Learn More
Medchemexpress LLC HY-B0546 500mg Medchemexpress, Procaine CAS:59-46-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0546500MG
Encompass_Preferred
Medchemexpress, HY-B0546 500mg Procaine CAS:59-46-1 Procaine is a DNA-demethylating agent. Procaine acts through multiple targets and has a slow onset and a short duration of action. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2589