Learn More
Medchemexpress LLC Procaine | 59-46-1 | 99.79% | 5 G
SDP
Catalog No. 5000408652
Encompass_Preferred
Procaine is a DNA-demethylating agent that works through multiple targets, characterized by a slow onset and short duration of action. It inhibits 5-HT3 receptor-mediated inward current and reduces 5-methylcytosine DNA content by 40%. Additionally, Procaine exhibits growth-inhibitory effects in cancer cells and acts as an excitant of limbic system cells, altering synaptic transmission.
  • Inhibits 5-HT3 receptor-mediated inward current
  • Reduces 5-methylcytosine DNA content
  • Demethylates densely hypermethylated CpG islands
  • Exhibits growth-inhibitory effects in cancer cells
  • Induces mitotic arrest
  • Acts as an excitant of limbic system cells
  • Alters synaptic transmission from the amygdala
Catalog No. 50-004-08652 Supplier Medchemexpress LLC Supplier No. HYB05465G
Procaine is a DNA-demethylating agent that works through multiple targets, characterized by a slow onset and short duration of action. It inhibits 5-HT3 receptor-mediated inward current and reduces 5-methylcytosine DNA content by 40%. Additionally, Procaine exhibits growth-inhibitory effects in cancer cells and acts as an excitant of limbic system cells, altering synaptic transmission.
  • Inhibits 5-HT3 receptor-mediated inward current
  • Reduces 5-methylcytosine DNA content
  • Demethylates densely hypermethylated CpG islands
  • Exhibits growth-inhibitory effects in cancer cells
  • Induces mitotic arrest
  • Acts as an excitant of limbic system cells
  • Alters synaptic transmission from the amygdala