Learn More
Apexbio Technology LLC PROCAINE-HCL-1G
SDP
Catalog No. 5000568058
Encompass_Preferred
Procaine HCl (CAS No 51-05-8) is a local anesthetic commonly used in biomedical research Originating from the synthesis of para-aminobenzoic acid and diethylaminoethanol it primarily functions by blocking sodium channels affecting nerve impulse transmission This results in the inhibition of depolarization thus providing its anesthetic effects In vitro activities are usually observed in the nanomolar to low micromolar range offering a versatile tool for probing cellular responses Procaine HCl is frequently applied in various cell models and animal studies often as a control in drug screening assays Its broad application spectrum includes evaluating nerve conductance and pain management mechanisms Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68058 Supplier Apexbio Technology LLC Supplier No. B22731000
Procaine HCl (CAS No 51-05-8) is a local anesthetic commonly used in biomedical research Originating from the synthesis of para-aminobenzoic acid and diethylaminoethanol it primarily functions by blocking sodium channels affecting nerve impulse transmission This results in the inhibition of depolarization thus providing its anesthetic effects In vitro activities are usually observed in the nanomolar to low micromolar range offering a versatile tool for probing cellular responses Procaine HCl is frequently applied in various cell models and animal studies often as a control in drug screening assays Its broad application spectrum includes evaluating nerve conductance and pain management mechanisms Experimental concentrations typically depend on the specific research design