Learn More
Medchemexpress LLC Procaine hydrochloride | 51-05-8 | 97.9% | 10 G
SDP
Catalog No. 5000408169
Encompass_Preferred
Procaine hydrochloride is a DNA-demethylating agent that acts through multiple targets and has a slow onset and a short duration of action. It is for research use only and not sold to patients. It functions as an excitant of limbic system cells and alters synaptic transmission in certain output pathways from the amygdale.
  • Inhibits the 5-HT3 receptor-mediated inward current.
  • Reduces 5-methylcytosine DNA content by 40%.
  • Demethylates densely hypermethylated CpG islands.
  • Exhibits growth-inhibitory effects in cancer cells, causing mitotic arrest.
Catalog No. 50-004-08169 Supplier Medchemexpress LLC Supplier No. HYB0546A10G
Procaine hydrochloride is a DNA-demethylating agent that acts through multiple targets and has a slow onset and a short duration of action. It is for research use only and not sold to patients. It functions as an excitant of limbic system cells and alters synaptic transmission in certain output pathways from the amygdale.
  • Inhibits the 5-HT3 receptor-mediated inward current.
  • Reduces 5-methylcytosine DNA content by 40%.
  • Demethylates densely hypermethylated CpG islands.
  • Exhibits growth-inhibitory effects in cancer cells, causing mitotic arrest.