Learn More
Medchemexpress LLC Procaine hydrochloride | 51-05-8 | 97.92% | 5 G
SDP
Catalog No. 5000408402
Encompass_Preferred
Procaine hydrochloride is a DNA-demethylating agent that functions through multiple targets. It is known for its slow onset and short duration of action, inhibiting 5-HT3 receptor-mediated inward current.
  • Reduces 5-methylcytosine DNA content by 40%
  • Can demethylate densely hypermethylated CpG islands
  • Exhibits growth-inhibitory effects in cancer cells, leading to mitotic arrest
  • Functions as an excitant of limbic system cells
  • Alters synaptic transmission in certain output pathways from the amygdale
Catalog No. 50-004-08402 Supplier Medchemexpress LLC Supplier No. HYB0546A5G
Procaine hydrochloride is a DNA-demethylating agent that functions through multiple targets. It is known for its slow onset and short duration of action, inhibiting 5-HT3 receptor-mediated inward current.
  • Reduces 5-methylcytosine DNA content by 40%
  • Can demethylate densely hypermethylated CpG islands
  • Exhibits growth-inhibitory effects in cancer cells, leading to mitotic arrest
  • Functions as an excitant of limbic system cells
  • Alters synaptic transmission in certain output pathways from the amygdale