Learn More
Medchemexpress LLC Procaine hydrochloride | 51-05-8 | 99.8% | 25 MG
SDP
Catalog No. 5000450777
Encompass_Preferred
Procaine hydrochloride (Standard) is the analytical standard of Procaine (hydrochloride), intended for research and analytical applications. It functions as a DNA-demethylating agent, acting through multiple targets with a slow onset and short duration of action. As an analytical grade compound, it serves as a reference standard and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • It is the analytical standard of procaine (hydrochloride).
  • The product is intended for research and analytical applications.
  • It is a DNA-demethylating agent.
  • It acts through multiple targets.
  • It has a slow onset and a short duration of action.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-50777 Supplier Medchemexpress LLC Supplier No. 25MGHYB0546AR
Procaine hydrochloride (Standard) is the analytical standard of Procaine (hydrochloride), intended for research and analytical applications. It functions as a DNA-demethylating agent, acting through multiple targets with a slow onset and short duration of action. As an analytical grade compound, it serves as a reference standard and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • It is the analytical standard of procaine (hydrochloride).
  • The product is intended for research and analytical applications.
  • It is a DNA-demethylating agent.
  • It acts through multiple targets.
  • It has a slow onset and a short duration of action.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.