Learn More
Medchemexpress LLC Procaine hydrochloride | 51-05-8 | MFCD00013000 | 99.8% | 272.77 | C13H21ClN2O2 | 10 MG
SDP
Catalog No. 5000298003
Encompass_Preferred
Procaine hydrochloride analytical standard provided as a 10 mg reference for research and analytical use. The hydrochloride salt of procaine, it is commonly used as an analytical reference and DNA-demethylating agent. The material is supplied with purity suitable for assay, method development, and quality control, along with lot-specific documentation.
Catalog No. 50-002-98003 Supplier Medchemexpress LLC Supplier No. HYB0546AR10MG
Procaine hydrochloride analytical standard provided as a 10 mg reference for research and analytical use. The hydrochloride salt of procaine, it is commonly used as an analytical reference and DNA-demethylating agent. The material is supplied with purity suitable for assay, method development, and quality control, along with lot-specific documentation.