Learn More
Medchemexpress LLC Procaine hydrochloride (Standard) | 51-05-8 | MFCD00013000 | 99.8% | 100 MG
SDP
Catalog No. 5000408642
Encompass_Preferred
Procaine hydrochloride (Standard) is an analytical standard and reference assay for research and analytical applications. It acts as a DNA-demethylating agent with a slow onset and short duration. This compound is suitable for qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications.
  • Acts as a DNA-demethylating agent.
  • Has a slow onset and short duration of action.
  • Suitable for qualitative research experiments.
  • Suitable for quantitative research experiments.
  • Suitable for methodological research experiments.
  • Used in HPLC.
  • Used in GC.
  • Used in MS.
Catalog No. 50-004-08642 Supplier Medchemexpress LLC Supplier No. HYB0546AR100MG
Procaine hydrochloride (Standard) is an analytical standard and reference assay for research and analytical applications. It acts as a DNA-demethylating agent with a slow onset and short duration. This compound is suitable for qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications.
  • Acts as a DNA-demethylating agent.
  • Has a slow onset and short duration of action.
  • Suitable for qualitative research experiments.
  • Suitable for quantitative research experiments.
  • Suitable for methodological research experiments.
  • Used in HPLC.
  • Used in GC.
  • Used in MS.