Learn More
Cusabio Technology LLC procalcitonin PCT
Catalog No. 5000051942
Encompass_Preferred
The Bovine Procalcitonin (PCT) ELISA Kit is designed for the quantitative detection of PCT in bovine serum plasma and tissue homogenates This kit detects PCT using a sandwich ELISA method providing a broad detection range of 39 06 pg/mL to 2500 pg/mL with a sensitivity of 9 77 pg/mL The assay requires a sample volume of 50-100 L and features an incubation time of 1-5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable and precise results for research or diagnostic applications The kit is optimized for accuracy and reproducibility making it suitable for studies involving inflammatory responses or infection monitoring in bovine samples
Catalog No. 50-000-51942 Supplier Cusabio Technology LLC Supplier No. CSBE15004B
The Bovine Procalcitonin (PCT) ELISA Kit is designed for the quantitative detection of PCT in bovine serum plasma and tissue homogenates This kit detects PCT using a sandwich ELISA method providing a broad detection range of 39 06 pg/mL to 2500 pg/mL with a sensitivity of 9 77 pg/mL The assay requires a sample volume of 50-100 L and features an incubation time of 1-5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable and precise results for research or diagnostic applications The kit is optimized for accuracy and reproducibility making it suitable for studies involving inflammatory responses or infection monitoring in bovine samples