Learn More
Selleck Chemical LLC Procarbazine HCl S1995-1g
SDP
Catalog No. 503111166
Encompass_Preferred
Procarbazine HCl (NSC-77213) is a hydrochloride salt form of procarbazine which is a polyfunctional alkylating compound used for the treatment of Hodgkin s lymphoma
Catalog No. 50-311-1166 Supplier Selleck Chemical LLC Supplier No. S19951G
Add to Cart
Add to Cart
Procarbazine HCl (NSC-77213) is a hydrochloride salt form of procarbazine which is a polyfunctional alkylating compound used for the treatment of Hodgkin s lymphoma