Learn More
Cayman Chemical Procarbazine (hydrochloride), Purity: ≥98%, CAS: 366-70-1, 100 mg
SDP
Catalog No. 502973184
Encompass_Preferred
A methyl hydrazine derivative that is converted by metabolism into a methylating species which converts DNA to various DNA adducts, including O6-methylguanine
Catalog No. 50-297-3184 Supplier Cayman Chemical Supplier No. 16133100 MG
A methyl hydrazine derivative that is converted by metabolism into a methylating species which converts DNA to various DNA adducts, including O6-methylguanine