Learn More
Cayman Chemical Procarbazine (hydrochloride), Purity: ≥98%, CAS: 366-70-1, 50 mg
SDP
Catalog No. 502970599
Encompass_Preferred
A methyl hydrazine derivative that is converted by metabolism into a methylating species which converts DNA to various DNA adducts, including O6-methylguanine
Catalog No. 50-297-0599 Supplier Cayman Chemical Supplier No. 1613350 MG
A methyl hydrazine derivative that is converted by metabolism into a methylating species which converts DNA to various DNA adducts, including O6-methylguanine