Learn More
Miele, Inc. ProCare Lab 10 AO - 5 L
Catalog No. 501713087
Encompass_Preferred
ProCare Lab 10 AO - 5 LLiquid detergent, alkaline, 5 l for optimum reprocessing of laboratory utensils.Note on the use of detergents with active chlorineOxidising effect, free from surfactantsSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 11.8-12.1
Catalog No. 50-171-3087 Supplier Miele, Inc. Supplier No. 10265580
ProCare Lab 10 AO - 5 LLiquid detergent, alkaline, 5 l for optimum reprocessing of laboratory utensils.Note on the use of detergents with active chlorineOxidising effect, free from surfactantsSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 11.8-12.1