Learn More
Miele, Inc. ProCare Lab 10 AP - 10L
Catalog No. 501713085
Encompass_Preferred
ProCare Lab 10 AP - 10LLiquid detergent, alkaline, 5 l for optimum reprocessing of laboratory utensils.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialspH value 12.3-12.5
Catalog No. 50-171-3085 Supplier Miele, Inc. Supplier No. 10265640
ProCare Lab 10 AP - 10LLiquid detergent, alkaline, 5 l for optimum reprocessing of laboratory utensils.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialspH value 12.3-12.5