Learn More
Miele, Inc. ProCare Lab 10 AP - 5L
Supplier:  Miele, Inc. 10265620
Encompass
ProCare Lab 10 AP - 5LLiquid detergent, alkaline, 5 l for optimum reprocessing of laboratory utensils.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialspH value 12.3-12.5
Catalog No. 50-171-3086