Learn More
Miele, Inc. ProCare Lab 10 AT - 5 L
Catalog No. 501713089
Encompass_Preferred
ProCare Lab 10 AT - 5 LLiquid detergent, alkaline, 5 l for optimum reprocessing of laboratory utensils.No phosphorous compounds & oxidants, contains surfactantsSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 12.2-12.5
Catalog No. 50-171-3089 Supplier Miele, Inc. Supplier No. 10265650
ProCare Lab 10 AT - 5 LLiquid detergent, alkaline, 5 l for optimum reprocessing of laboratory utensils.No phosphorous compounds & oxidants, contains surfactantsSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 12.2-12.5