Learn More
Miele, Inc. ProCare Lab 10 MA - 10 L
Catalog No. 501713090
Encompass_Preferred
ProCare Lab 10 MA - 10 LLiquid detergent, mildly alkaline, 10 l for optimum reprocessing of laboratory glassware and utensils.Free from surfactants and oxidantsSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 11.6-11.8
Catalog No. 50-171-3090 Supplier Miele, Inc. Supplier No. 10265680
ProCare Lab 10 MA - 10 LLiquid detergent, mildly alkaline, 10 l for optimum reprocessing of laboratory glassware and utensils.Free from surfactants and oxidantsSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 11.6-11.8