Learn More
Miele, Inc. ProCare Lab 11 AP - 10 kg
Catalog No. 501713092
Encompass_Preferred
ProCare Lab 11 AP - 10 kgPowder cleaning agent, alkaline, 10 kg for optimum reprocessing of laboratory utensils.Free from phosphorous compounds, oxidising effectSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 11.5-11.7
Catalog No. 50-171-3092 Supplier Miele, Inc. Supplier No. 10266540
ProCare Lab 11 AP - 10 kgPowder cleaning agent, alkaline, 10 kg for optimum reprocessing of laboratory utensils.Free from phosphorous compounds, oxidising effectSpecially developed for Miele ProfessionalSuitable for all types of materialsPH value 11.5-11.7