Learn More
Miele, Inc. ProCare Lab 30 C - 5 L
Catalog No. 501713095
Encompass_Preferred
ProCare Lab 30 C - 5 LNeutralizing agent, acidic, 5 l for optimum neutralization with citric acid based neutralizing agent.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialsTo be used as neutralizing agent or acid pre-cleanerpH value 2.6-3
This item is not returnable. View return policy
Catalog No. 50-171-3095 Supplier Miele, Inc. Supplier No. 10266670
ProCare Lab 30 C - 5 LNeutralizing agent, acidic, 5 l for optimum neutralization with citric acid based neutralizing agent.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialsTo be used as neutralizing agent or acid pre-cleanerpH value 2.6-3