Learn More
Miele, Inc. ProCare Lab 30 P - 5L
Catalog No. 501713097
Encompass_Preferred
ProCare Lab 30 P - 5LNeutralizing agent, acidic, 5 l for optimum neutralization with citric acid based neutralizing agent.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialsTo be used as neutralizing agent or acid pre-cleanerpH value 2.6-3
Catalog No. 50-171-3097 Supplier Miele, Inc. Supplier No. 10266490
ProCare Lab 30 P - 5LNeutralizing agent, acidic, 5 l for optimum neutralization with citric acid based neutralizing agent.Free from surfactants, phosphorus compounds, and oxidation agentsSpecially developed for Miele ProfessionalSuitable for all types of materialsTo be used as neutralizing agent or acid pre-cleanerpH value 2.6-3