Learn More
Miele, Inc. ProCare Universal 61 - 6 kg
Catalog No. 501713117
Encompass_Preferred
ProCare Universal 61 - 6 kgRegeneration salt, 3 x 2 kg (13.2 lb total) for regeneration of the integrated water softener.Prevents calcium deposits and protects wash items
Catalog No. 50-171-3117 Supplier Miele, Inc. Supplier No. 09195780
ProCare Universal 61 - 6 kgRegeneration salt, 3 x 2 kg (13.2 lb total) for regeneration of the integrated water softener.Prevents calcium deposits and protects wash items