Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProcartaPlex™ Cell Lysate Assay Diluent (1X)

Catalog No. 502557502
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-7502 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-7502 Supplier Invitrogen™ Supplier No. EPX11114000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ProcartaPlex Cell Lysate Assay Diluent (1X) is for use with the Apoptosis 6-Plex Human ProcartaPlex Panel for dilution of cell lysate samples.

ProcartaPlex Cell Lysate Assay Diluent (1X) is for use with the Apoptosis 6-Plex Human ProcartaPlex Panel (Cat. No. EPX060-15823-901) for dilution of cell lysate samples. Diluent is included with the panel, but is also offered here as a stand-alone item.

Reported application

  • Multiplex immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Product Type Diluent
Quantity 10 mL
Product Line ProcartaPlex
Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.