Learn More
Invitrogen™ ProcartaPlex™ Flat Bottom Plate, 96-well
Catalog No. 502557498
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
Catalog No. Quantity
50-255-7498 96 tests
1 options
Catalog No. 50-255-7498 Supplier Invitrogen™ Supplier No. EPX88182000
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart
This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.
This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.
This plate is included in all ProcartaPlex multiplex panel kits, species-specific basic kits, and as part of custom mix and match panels. For bulk orders, please contact your sales representative.
Note that the reusable lid (Cat. No. EPX-88104-000) is sold separately.
About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to detect and quantitate up to 80 protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, non-human primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.
Related products
  • ProcartaPlex Microplate Lid (Cat. No. EPX-88104-000)
TRUSTED_SUSTAINABILITY
Specifications
Lid Without Lid
Product Line ProcartaPlex
Color Black
Disposable Single-use
Product Type Plate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Hydrophobic
Well Shape Flat Bottom (F-well)
Quantity 96 tests
Fit for Purpose Certified Quality