Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAPK Pathway Panel (phospho), 6plex

Catalog No. 502884750
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-288-4750 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-288-4750 Supplier Invitrogen™ Supplier No. EPX06097117PHO
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen ProcartaPlex Human MAPK Pathway Panel (phospho), 6plex enables precise and simultaneous analysis of 6 phosphorylated proteins involved in the MAPK pathway.

The ProcartaPlex Human MAPK Pathway Panel (phospho), 6plex, enables simultaneous quantitation of six phosphorylated proteins included in the MAPK cell signaling pathway. It complements the ProcartaPlex Human MAPK Pathway Panel (total) 6plex (Cat. No. EPX060-97017-901) that measures the total protein level of the same six proteins.

The panel is validated for use with cell lysates. For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single to Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included in the panel. Depending on your cell system, other buffers can be suitable as well.

This kit is compatible with all Luminex instruments. If you use an xMAP INTELLIFLEX DR-SE instrument, we recommend our ProcartaPlex Human MAPK Pathway Dual Reporter Panel 2x6plex (Cat. No. EPX060-97217-DR), which enables the simultaneous quantitation of total and phosphorylated proteins on the same bead in a single well.

The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

Targets [bead region]: JNK1/3[pT183/pT221] [13], p38[pT180/pY182] [14], FAK[pY397] [30], c-Myc[pS62] [53], ERK1[pT202/pY204] [62], MEK1[pS218/pS222] [63]

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample - measure multiple protein targets simultaneously in a single 25 to 50 μL sample
  • Well-established Luminex technology - highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25 to 50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin to R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Type Magnetic
Bead Region JNK1/3[pT183/pT221] [13], p38[pT180/pY182] [14], FAK[pY397] [30], c-Myc[pS62] [53], ERK1[pT202/pY204] [62], MEK1[pS218/pS222] [63]
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
For Use With (Equipment) Luminex™ Instruments
High-throughput Compatibility Yes
Interassay CV See Certificate of Analysis
Intraassay CV See Certificate of Analysis
Multiplex Size Plex No. 6plex
Product Line ProcartaPlex
Sample Type Cell lysates
Sample Volume Cell lysates (prediluted): 25 μL
Content And Storage 2 vials Standard Mix (lyophilized)
1 vial Detection Antibody Mix (1X)
1 vial Bead Mix (1X)
1 bottle Universal Assay Buffer (10X)
1 bottle Streptavidin-PE (SA-PE) (1X)
1 bottle Wash Buffer (10X)
2 8-Tube Strip
1 Flat Bottom 96-well Plate, black
1 Microplate Lid
Plate Seals

Store as indicated on the Certificate of Analysis.
Shipping Condition Wet Ice
Research Area Signaling
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.