Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEK1[pS218/pS222] Human ProcartaPlex™ Simplex Kit

Catalog No. 502884748
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-288-4748 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-288-4748 Supplier Invitrogen™ Supplier No. EPX01097114PHO
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen ProcartaPlex Human MEK1[pS218/pS222] Simplex Kit enables precise analysis of the Protein of Interest.

The ProcartaPlex Human MEK1[pS218/pS222] Simplex Kit measures the level of phosphorylated MEK1[pS218/pS222] in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single to Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included in the kit. Depending on your cell system, other buffers can be suitable as well.

The ProcartaPlex Human MEK1[pS218/pS222] Simplex Kit is part of the ProcartaPlex Human MAPK Pathway Panel (phospho) 6plex (Cat. No. EPX060-97117-PHO), which complements the ProcartaPlex Human MAPK Pathway Panel (total) 6plex (Cat. No. EPX060-97017-901).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Type Magnetic
Bead Region MEK1[pS218/pS222] [63]
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
For Use With (Equipment) Luminex™ Instruments
Gene MAP2K1
Gene Alias 5604
Gene ID (Entrez) MAP2K1
High-throughput Compatibility Yes
Interassay CV See Certificate of Analysis
Intraassay CV See Certificate of Analysis
Product Line ProcartaPlex
Sample Type Cell lysates
Sample Volume Cell lysates (prediluted): 25 μL
UniProt ID Q02750
Synonym Mitogen-activated protein kinase
Content And Storage 1 vial Capture Beads (50x)
1 vial biotinylated Detection Antibody (50x)
2 vials Standard Mix (lyophilized)

Store as indicated on the Certificate of Analysis.
Shipping Condition Wet Ice
Research Area Signaling
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.