Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NF-L Human ProcartaPlex™ Simplex Kit

Catalog No. 5000031036
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-000-31036 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-000-31036 Supplier Invitrogen™ Supplier No. EPX01012362901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen ProcartaPlex Human NF-L Simplex kit enables precise analysis of the Protein of Interest.

The human NF-L ProcartaPlex Simplex Kit measures NF-L protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Type Magnetic
Bead Region NF-L [78]
Assay Sensitivity 5.9 pg/mL
Assay Range 18-75000 pg/mL
For Use With (Equipment) Luminex™ Instruments
Gene NEFL
Gene Alias Neurofilament Light Chain, NF68 2 3 4 5 NFL, CMT1F, CMT2E, Protein Phosphatase, Regulatory Subunit Neurofilament, Light Polypeptide 68kDa
Gene ID (Entrez) 4747
Gene Symbol NEFL
High-throughput Compatibility Yes
Interassay CV 5%
Intraassay CV <10%
Product Line ProcartaPlex
Sample Type Serum, Plasma, CSF
Sample Volume 25 μL
UniProt ID P07196
Synonym Neurofilament Light Chain, NEFL
Content And Storage 1 vial Capture Beads (50x)
1vial biotinylated Detection Antibody (50x)
2 vials Standard Mix (lyophilized)

Store as indicated on the Certificate of Analysis.
Shipping Condition Wet Ice
Research Area Neurobiology
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.