Learn More
Invitrogen™ ProcartaPlex™ Microplate Lid
Catalog No. 502557499
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Catalog No. Color
50-255-7499 Black
1 options
Catalog No. 50-255-7499 Supplier Invitrogen™ Supplier No. EPX88104000
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart
This reusable microplate lid is ideal for use with all ProcartaPlex Luminex protein assays.
This reusable microplate lid is ideal for use with all ProcartaPlex Luminex protein assays.
This lid is included in all ProcartaPlex multiplex panel kits, species-specific basic kits, and as part of custom mix and match panels. For bulk orders, please contact your sales representative.
Note that the 96-well flat bottom plate (Cat. No. EPX-88182-000) is sold separately.
About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to detect and quantitate up to 80 protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, non-human primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.
Related products
  • ProcartaPlex Flat Bottom Plate, 96-well (Cat. No. EPX-88182-000)
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polystyrene
Sterility Non-sterile
No. of Wells 96
No. per Pack 1
Quantity 1
Product Type Plate
Fit for Purpose Certified Quality