Learn More
Invitrogen™ Mouse Detection Antibody Diluent
Description
The ProcartaPlex Detection Antibody Diluent is used for dilution of detection antibodies.
The ProcartaPlex Detection Antibody Diluent is included in our basic kits and off the shelf panels. Additionally, we offer the Detection Antibody Diluent as a stand-alone item.
Reported Application
Multiplex Immunoassay
Specifications
Specifications
| Product Type | Diluent |
| Quantity | 1 each |
| Species | Mouse |
| For Use With (Equipment) | Luminex™ Instruments |
Frequently Asked Questions (FAQs)
No, the standalone ProcartaPlex assay buffers are not lot-specific.
Here are the catalog numbers:
- EPX-11111-000: ProcartaPlex Universal Assay Buffer (1X) (10 mL) - It replaces all discontinued Procarta buffers (standard, assay, sample buffer); it is the same as the discontinued Procarta Cell Lysate Assay Buffer, 10 plate (Cat. No. PC0165), but only for one plate
- EPX-11110-000: ProcartaPlex Universal Assay Buffer (10X) (10 mL)
- EPXP-11112-000: ProcartaPlex Platinum Assay Buffer for Plasma Samples (1X, 10 mL)
- EPXP-11113-000: ProcartaPlex Platinum Assay Buffer for Serum Samples (1X, 10 mL)
- EPX-99999-000: ProcartaPlex Cell Lysis Buffer; It is the same as the discontinued Procarta Cell lysis Buffer (for cell lysate preparation) (Cat. No. PC6002)
- EPX-22222-000: ProcartaPlex Detection Antibody Diluent, Mouse (1X, 3 mL)
- EPX-33333-000: ProcartaPlex Detection Antibody Diluent, Human (1X, 3 mL)
- EPX-66666-000: ProcartaPlex Wash Buffer 10X (20 mL)
- EPX-66666-001: ProcartaPlex Wash Buffer 10X (200 mL)
- EPX-77777-000: ProcartaPlex Reading Buffer (1X, 40 mL)
- EPX-SAPE-000: ProcartaPlex Streptavidin-PE (1X, 5 mL) - 4.3 µg/mL (can range from 4.25-4.35 µg/mL)
- EPX-44444-000: ProcartaPlex 96 well Flat Bottom Plate, black (1 plate)
- EPX-88888-000: ProcartaPlex 96 well Filter Plate (1 plate)
- EPX-38488-000: ProcartaPlex 384 well Filter Plate (1 plate)
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.