Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProcartaPlex™ NHP Control Set, 37plex

Catalog No. 502556990
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-6990 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-6990 Supplier Invitrogen™ Supplier No. EPX37040045CTR
Add to Cart
Add to Cart

The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel.

The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel. For a list of analytes included in this set, please refer to the TDS in the Documents section.

Reported application

  • Multiplex Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Control Set
Quantity 1 set
Species Non-Human Primate
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Content And Storage

• 1 vial NHP Control High A
• 1 vial NHP Control High B
• 1 vial NHP Control Low A
• 1 vial NHP Control Low B

Store at 2°C to 8°C.

Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.