Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProcartaPlex™ Platinum Assay Buffer for Plasma Samples

Catalog No. 502556985
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-6985 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-6985 Supplier Invitrogen™ Supplier No. EPXP11112000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ProcartaPlex Platinum Assay Buffer for Plasma Samples is used for dilution of plasma samples and gives spike- and dilution-recovery results in the range of 70 to 130%.

ProcartaPlex Platinum Assay Buffer for Plasma Samples is used for dilution of plasma samples and gives spike- and dilution-recovery results in the range of 70–130%.

ProcartaPlex Platinum Assay Buffer for Plasma Samples is included in our Platinum ProcartaPlex Human Basic Kit for Plasma Samples (Cat.No. EPXP010-10421-901) and off-the-shelf Platinum panels. It is offered here as a stand-alone item.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Buffer
Quantity 10 mL
Species Human
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Sample Type Plasma
Content And Storage Store in refrigerator 2°C to 8°C
Shipping Condition Wet Ice
What species can ProcartaPlex Platinum Assay Buffer for Plasma Samples be used with?

ProcartaPlex Platinum Assay Buffer for Plasma Samples is designed to be used with the ProcartaPlex Platinum assay format. Currently, this format has been validated for human samples only. Other species have not been tested.

What kits can ProcartaPlex Platinum Assay Buffer for Plasma Samples be used with?

ProcartaPlex Platinum Assay Buffer for Plasma Samples is designed to be used with the ProcartaPlex Platinum assay format. It is used for dilution of plasma samples and gives spike- and dilution-recovery results in the range of 70-130%. ProcartaPlex Platinum Assay Buffer for Plasma Samples is included in our Platinum ProcartaPlex Human Basic Kit for Plasma Samples (Cat.No. EPXP010-10421-901) and off-the-shelf Platinum panels. It is also offered as a stand-alone item (Cat. No. EPXP-11112-000).

Are the standalone ProcartaPlex assay buffers lot-specific such that they will only work with specific lots of other components?

No, the standalone ProcartaPlex assay buffers are not lot-specific.

Can I get the catalog numbers for ProcartaPlex assay buffers and plates?

Here are the catalog numbers:

- EPX-11111-000: ProcartaPlex Universal Assay Buffer (1X) (10 mL) - It replaces all discontinued Procarta buffers (standard, assay, sample buffer); it is the same as the discontinued Procarta Cell Lysate Assay Buffer, 10 plate (Cat. No. PC0165), but only for one plate
- EPX-11110-000: ProcartaPlex Universal Assay Buffer (10X) (10 mL)
- EPXP-11112-000: ProcartaPlex Platinum Assay Buffer for Plasma Samples (1X, 10 mL)
- EPXP-11113-000: ProcartaPlex Platinum Assay Buffer for Serum Samples (1X, 10 mL)
- EPX-99999-000: ProcartaPlex Cell Lysis Buffer; It is the same as the discontinued Procarta Cell lysis Buffer (for cell lysate preparation) (Cat. No. PC6002)
- EPX-22222-000: ProcartaPlex Detection Antibody Diluent, Mouse (1X, 3 mL)
- EPX-33333-000: ProcartaPlex Detection Antibody Diluent, Human (1X, 3 mL)
- EPX-66666-000: ProcartaPlex Wash Buffer 10X (20 mL)
- EPX-66666-001: ProcartaPlex Wash Buffer 10X (200 mL)
- EPX-77777-000: ProcartaPlex Reading Buffer (1X, 40 mL)
- EPX-SAPE-000: ProcartaPlex Streptavidin-PE (1X, 5 mL) - 4.3 µg/mL (can range from 4.25-4.35 µg/mL)
- EPX-44444-000: ProcartaPlex 96 well Flat Bottom Plate, black (1 plate)
- EPX-88888-000: ProcartaPlex 96 well Filter Plate (1 plate)
- EPX-38488-000: ProcartaPlex 384 well Filter Plate (1 plate)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.