Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProcartaPlex™ Wash Buffer (10X)

Catalog No. EPX66666001
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EPX66666001 200 mL
50-112-5865 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EPX66666001 Supplier Invitrogen™ Supplier No. EPX66666001
Add to Cart
Add to Cart

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates.

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates. It is a simple mix of buffers, salts, and mild detergent.

ProcartaPlex Wash Buffer (10X) is included in our basic kits and off-the-shelf panels. Additionally, we offer it here as a stand-alone item.

Reported application

  • Multiplex immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Product Type Buffer
Quantity 200 mL
For Use With (Equipment) Luminex™ Instruments
Are the standalone ProcartaPlex assay buffers lot-specific such that they will only work with specific lots of other components?

No, the standalone ProcartaPlex assay buffers are not lot-specific.

At what temperature and for how long can the diluted ProcartaPlex Wash Buffer be stored?

The diluted buffer can be stored at 4 degrees C and should be used within 1 month of diluting.

Can I get the catalog numbers for ProcartaPlex assay buffers and plates?

Here are the catalog numbers:

- EPX-11111-000: ProcartaPlex Universal Assay Buffer (1X) (10 mL) - It replaces all discontinued Procarta buffers (standard, assay, sample buffer); it is the same as the discontinued Procarta Cell Lysate Assay Buffer, 10 plate (Cat. No. PC0165), but only for one plate
- EPX-11110-000: ProcartaPlex Universal Assay Buffer (10X) (10 mL)
- EPXP-11112-000: ProcartaPlex Platinum Assay Buffer for Plasma Samples (1X, 10 mL)
- EPXP-11113-000: ProcartaPlex Platinum Assay Buffer for Serum Samples (1X, 10 mL)
- EPX-99999-000: ProcartaPlex Cell Lysis Buffer; It is the same as the discontinued Procarta Cell lysis Buffer (for cell lysate preparation) (Cat. No. PC6002)
- EPX-22222-000: ProcartaPlex Detection Antibody Diluent, Mouse (1X, 3 mL)
- EPX-33333-000: ProcartaPlex Detection Antibody Diluent, Human (1X, 3 mL)
- EPX-66666-000: ProcartaPlex Wash Buffer 10X (20 mL)
- EPX-66666-001: ProcartaPlex Wash Buffer 10X (200 mL)
- EPX-77777-000: ProcartaPlex Reading Buffer (1X, 40 mL)
- EPX-SAPE-000: ProcartaPlex Streptavidin-PE (1X, 5 mL) - 4.3 µg/mL (can range from 4.25-4.35 µg/mL)
- EPX-44444-000: ProcartaPlex 96 well Flat Bottom Plate, black (1 plate)
- EPX-88888-000: ProcartaPlex 96 well Filter Plate (1 plate)
- EPX-38488-000: ProcartaPlex 384 well Filter Plate (1 plate)

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.