Learn More
Selleck Chemical LLC Procaterol HCl S2083-5mg
SDP
Catalog No. 503110019
Encompass_Preferred
Procaterol HCl (OPC-2009) is a short-acting 2-adrenergic receptor agonist with Kp of 8 nM used for the treatment of asthma
Catalog No. 50-311-0019 Supplier Selleck Chemical LLC Supplier No. S20835MG
Add to Cart
Add to Cart
Procaterol HCl (OPC-2009) is a short-acting 2-adrenergic receptor agonist with Kp of 8 nM used for the treatment of asthma