Learn More
Medchemexpress LLC Procaterol (hydrochloride hemihydrate) | 81262-93-3 | 99.8% | 25 MG
SDP
Catalog No. 5000343042
Encompass_Preferred
Procaterol hydrochloride hemihydrate is an orally active β2 adrenoreceptor agonist primarily used for asthma research. It demonstrates significant biological activity in both in vitro and in vivo studies, including reducing rhinovirus infection, protecting cells from oxidative stress, and inhibiting inflammatory responses. This compound also alleviates cough in animal models, making it a valuable tool for related scientific investigations.
  • Orally active β2 adrenoreceptor agonist
  • Utilized for asthma research
  • Decreases rhinovirus infection in human tracheal cells
  • Inhibits IL-1β- and TNF-α-mediated eosinophil chemotactic activity
  • Protects cells from H2O2-induced oxidative stress
  • Inhibits histamine-induced airflow obstruction and microvascular leakage
  • Abolishes Mch-induced cough in guinea pigs
Catalog No. 50-003-43042 Supplier Medchemexpress LLC Supplier No. 25MGHY129041
Procaterol hydrochloride hemihydrate is an orally active β2 adrenoreceptor agonist primarily used for asthma research. It demonstrates significant biological activity in both in vitro and in vivo studies, including reducing rhinovirus infection, protecting cells from oxidative stress, and inhibiting inflammatory responses. This compound also alleviates cough in animal models, making it a valuable tool for related scientific investigations.
  • Orally active β2 adrenoreceptor agonist
  • Utilized for asthma research
  • Decreases rhinovirus infection in human tracheal cells
  • Inhibits IL-1β- and TNF-α-mediated eosinophil chemotactic activity
  • Protects cells from H2O2-induced oxidative stress
  • Inhibits histamine-induced airflow obstruction and microvascular leakage
  • Abolishes Mch-induced cough in guinea pigs