Learn More
Enzo Life Sciences Procathepsin K (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3670010
Encompass_Preferred
Alternative name: Procathepsin O, Procathepsin O2, Procathepsin X. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 27.8µl 20mM sodium phosphate, pH 6.9, 500mM NaCl, 1 mM EDTA. Source: Produced in insect cells. Full length glycosylated human procathepsin K (also known as cathepsin O or cathepsin O2) cloned from human cDNA. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2002