Learn More
Enzo Life Sciences Procathepsin V (human), (recombinant) (His-tag) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5540010
Encompass_Preferred
Full-length proenzyme. Can be activated in one step. Full-length proenzyme. Can be activated in one step. Alternative name: Cathepsin L2. Purity: ≥90%. Formulation: Liquid. In 25mM TRIS-HCl, pH 8.0, containing 100mM sodium chloride, 0.05% Tween-20 and 10% glycerol. Source: Produced in insect cells. Recombinant glycosylated procathepsin V cloned from human cDNA and purified as full-length proenzyme. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2087