Learn More
Biotang Inc MS-H-S Analog Magnetic Hotplate Stirrer, porcelain plate with Heating, speed range 0-1500 rpm, Temperature range: Ambient to 350deg.C (662 deg.F), plate diameter 135 mm
Catalog No. 507514912 Shop All BIOTANG Inc Products
Encompass_Preferred
MS-H-S Analog Magnetic Hotplate Stirrer, porcelain plate with Heating, speed range 0-1500 rpm, Temperature range: Ambient to 350deg.C (662 deg.F), plate diameter 135 mm
Catalog No. 50-751-4912 Supplier Biotang Inc Supplier No. M10008
MS-H-S Analog Magnetic Hotplate Stirrer, porcelain plate with Heating, speed range 0-1500 rpm, Temperature range: Ambient to 350deg.C (662 deg.F), plate diameter 135 mm