Learn More
Microsolv Technology Corporation Process HPLC Column, RP C18, Wide Pore, 10um, 300A. 50mm ID x 50mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68018P50503M
Encompass_Preferred
This Cogent RP C18™ Wide Pore, Stationary Phase has been a premium value, and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity 300Å silica available. The 10 µm Particle works well with higher flow rates and produces lower back pressures than smaller particles. The extra large diameter of this column provides high loading capacity which is great for purification of large compounds making it ideal for Process HPLC of many proteins & peptides and other compounds. A specialized HPLC System is required for Columns this size.
Catalog No. 50-226-9827
Add to Cart
Request a Quote