Learn More
Microsolv Technology Corporation Process HPLC Column, RP C18, Wide Pore, 5um, 300A. 50mm ID x 100mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68518P501003M
Encompass_Preferred
This Cogent RP C18™ Wide Pore, Stationary Phase has been a premium value, and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity 300Å silica available. The 5 µm Particle works well with higher flow rates and produces lower back pressures than smaller particles. The extra large diameter and long length of this column provides very high loading capacity which is great for purification of large compounds making it ideal for Process HPLC of many proteins & peptides and other compounds. A specialized HPLC System is required for Columns this size
Catalog No. 50-226-9880
Add to Cart
Request a Quote